Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A408997-1G A408997-5G A408997-100MG A408997-250MG A408997-25G
103827-194EA 86 USD InStock
103827-194 103838-242 77004-016 77004-018 77882-079
2-Bromo-6-nitropyridine 97%
2-Bromo-6-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00234012
CAS Number: 21203-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty