Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-9-phenylcarbazole 98%

Supplier: AMBEED, INC
Warning

A119061-1G A119061-100G A119061-10G A119061-250MG A119061-25G A119061-1G A119061-5G
103799-158EA 6 USD InStock
103799-158 76983-114 76983-116 76983-118 76983-120 77311-824 77311-826
2-Bromo-9-phenylcarbazole 98%
2-Bromo-9-phenylcarbazole
Precaución: for Research Use Only


Formula: C₁₈H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD24387061
CAS Number: 94994-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty