Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-bromopyridine 98%

Supplier: AMBEED, INC
Warning

A171657-1G A171657-1G A171657-25G A171657-5G A171657-100G A171657-10G A171657-500G
103806-654EA 8 USD InStock
103806-654 103844-930 77082-096 77082-098 77082-100 77286-890 77286-892 77286-894
3-Amino-2-bromopyridine 98%
3-Amino-2-bromopyridine
Precaución: for Research Use Only


Formula: C₅H₅BrN₂
Boiling Pt: 292.5 °C (760 mm Hg)
Melting Pt: 76…80 °C
Flash Pt: 130.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00234064
CAS Number: 39856-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty