Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-bromopyridine 98%

Supplier: AMBEED, INC
Danger

A178572-1G A178572-5G A178572-1G A178572-25G A178572-5G A178572-100G A178572-10G A178572-250MG
103807-356EA 6 USD InStock
103807-356 103845-692 77083-124 77083-126 77083-128 77273-964 77273-966 77273-968
4-Amino-2-bromopyridine 98%
4-Amino-2-bromopyridine
Precaución: for Research Use Only


Formula: C₅H₅BrN₂
MW: 173.01
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD01646061
CAS Number: 7598-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty