Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoterephthalic acid 98%

Supplier: AMBEED, INC
Warning

A542628-25G A542628-5G A542628-100G A542628-500G
103839-764EA 10 USD InStock
103839-764 76843-816 76843-818 77197-418 77197-420
2-Bromoterephthalic acid 98%
2-Bromoterephthalic acid
Precaución: for Research Use Only


Formula: C₈H₅BrO₄
MW: 245.03
Boiling Pt: 421.6 °C (760 mm Hg)
Melting Pt: 297…300 °C
Flash Pt: 208.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002403
CAS Number: 586-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty