Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiazole 98%

Supplier: AMBEED, INC
Warning

A192763-10G A192763-500G A192763-5G A192763-25G A192763-1000G
103809-278EA 147 USD InStock
103809-278 76974-920 76974-922 76974-924 77084-970 77302-320
2-Bromothiazole 98%
2-Bromothiazole
Precaución: For research use only.


Formula: C₃H₂BrNS
Boiling Pt: 171 °C (760 mm Hg)
Flash Pt: 67 °C
Storage Temperature: Freezer
MDL Number: MFCD00005316
CAS Number: 3034-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty