Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-thiazolecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A103142-1G A103142-5G A103142-100G A103142-10G A103142-250MG A103142-25G A103142-1G
103796-488EA 7 USD InStock
103796-488 103822-192 76809-376 76809-378 76809-380 76809-382 77309-066
2-Bromo-4-thiazolecarboxylic acid 97%
2-Bromo-4-thiazolecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₂BrNO₂S
MW: 208.04
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 212…214 °C
Flash Pt: 165.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115729
CAS Number: 5198-88-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty