Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-hydroxymethylthiazole 97%

Supplier: AMBEED, INC
Warning

A183082-1G A183082-5G A183082-1G A183082-5G A183082-250MG
103815-602EA 16 USD InStock
103815-602 103846-166 77083-750 77083-752 77163-934
2-Bromo-5-hydroxymethylthiazole 97%
2-Bromo-5-hydroxymethylthiazole
Precaución: for Research Use Only


Formula: C₄H₄BrNOS
Boiling Pt: 288.3 °C (760 mm Hg)
Flash Pt: 128.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04115733
CAS Number: 687636-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty