Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butynoic acid 98%

Supplier: AMBEED, INC
Danger

A717241-5G A717241-1G A717241-5G A717241-10G A717241-100G
103840-718EA 12 USD InStock
103840-718 76844-934 76844-936 76879-780 77673-970
2-Butynoic acid 98%
2-Butynoic acid
Precaución: for Research Use Only


Formula: C₄H₄O₂
MW: 84.07
Boiling Pt: 203
Melting Pt: 78
Density: 0.9641
Flash Pt: 90.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004363
CAS Number: 590-93-2
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty