Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1,10-phenanthroline 95%

Supplier: AMBEED, INC
Warning

A182512-1G A182512-1G A182512-5G A182512-250MG A182512-100G
103807-722EA 8 USD InStock
103807-722 103846-094 77083-640 77083-642 77260-290 77748-190
2-Chloro-1,10-phenanthroline 95%
2-Chloro-1,10-phenanthroline
Precaución: For research use only.


Formula: C₁₂H₇ClN₂
MW: 214.65 g/mol
Boiling Pt: 391.8 °C (760 mm Hg)
Melting Pt: 151…152 °C
Flash Pt: 222.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185678
CAS Number: 7089-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty