Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1,8-naphthyridine 96%

Supplier: AMBEED, INC
Warning

A250884-1G A250884-100MG A250884-250MG A250884-1G
103811-956EA 100 USD InStock
103811-956 77261-988 77261-990 77304-540
2-Chloro-1,8-naphthyridine 96%
2-Chloro-1,8-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₅ClN₂
Boiling Pt: 281.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00234390
CAS Number: 15936-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty