Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-α-Chlorophenylacetic acid 96%

Supplier: AMBEED, INC
Warning

A103105-1G A103105-5G A103105-100MG A103105-250MG A103105-1G
103796-482EA 15 USD InStock
103796-482 103822-186 76687-106 76687-108 77283-034
(±)-α-Chlorophenylacetic acid 96%
(±)-α-Chlorophenylacetic acid
Precaución: For research use only.


Formula: C₈H₇ClO₂
Boiling Pt: 282 °C (760 mm Hg)
Flash Pt: 124.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037753
CAS Number: 4755-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty