Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-fluoroaniline 98%

Supplier: AMBEED, INC
Warning

A104991-1G A104991-5G A104991-100G A104991-10G A104991-25G A104991-1G A104991-5G
103796-810EA 5 USD InStock
103796-810 103822-538 77283-318 77283-320 77283-322 77309-402 77309-404
2-Chloro-3-fluoroaniline 98%
2-Chloro-3-fluoroaniline
Precaución: for Research Use Only


Formula: C₆H₅ClFN
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03407961
CAS Number: 21397-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty