Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-formylpyridine 98%

Supplier: AMBEED, INC
Warning

A176701-1G A176701-100G A176701-10G A176701-500G A176701-1G A176701-25G A176701-5G
103807-166EA 9 USD InStock
103807-166 103845-492 76974-266 76974-268 76974-270 77082-840 77082-842 77082-844
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine
Precaución: for Research Use Only


Formula: C₆H₄ClNO
MW: 141.56
Boiling Pt: 242.2 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01315308
CAS Number: 36404-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty