Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-nitrothiophene 95%

Supplier: AMBEED, INC
Warning

A163788-1G A163788-5G A163788-250MG A163788-1G A163788-5G
103805-768EA 45 USD InStock
103805-768 103844-032 76812-902 77320-144 77320-146
2-Chloro-3-nitrothiophene 95%
2-Chloro-3-nitrothiophene
Precaución: for Research Use Only


Formula: C₄H₂ClNO₂S
Boiling Pt: 253.5 °C (760 mm Hg)
Melting Pt: 51 °C
Flash Pt: 107.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052159
CAS Number: 5330-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty