Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-iodopyrimidine 95%

Supplier: AMBEED, INC
Warning

A100697-1G A100697-5G A100697-100MG A100697-250MG A100697-5G
103795-582EA 201 USD InStock
103795-582 103821-278 76982-146 76982-148 77296-818
2-Chloro-4-iodopyrimidine 95%
2-Chloro-4-iodopyrimidine
Precaución: for Research Use Only


Formula: C₄H₂ClIN₂
MW: 240.43 g/mol
Boiling Pt: 317.8±15.0 °C (760 mm Hg)
Melting Pt: 111.3…112.4 °C
Flash Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18415735
CAS Number: 395082-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty