Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-nitrobenzene-1-sulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A741899-5G A741899-100MG A741899-250MG
103840-826EA 324 USD InStock
103840-826 77267-794 77267-796
2-Chloro-4-nitrobenzene-1-sulfonyl chloride 95%
2-Chloro-4-nitrobenzene-1-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO₄S
Boiling Pt: 370.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03954008
CAS Number: 20201-03-0
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty