Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-phenylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A157362-1G A157362-5G A157362-10G A157362-250MG A157362-1G
103804-992EA 35 USD InStock
103804-992 103843-194 77259-486 77259-488 77319-010
2-Chloro-4-phenylpyrimidine 98%
2-Chloro-4-phenylpyrimidine
Precaución: for Research Use Only


Formula: C₁₀H₇ClN₂
Boiling Pt: 368.3 °C (760 mm Hg)
Melting Pt: 86 °C
Flash Pt: 208.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234600
CAS Number: 13036-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty