Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(trifluoromethyl)nicotinic acid 98%

Supplier: AMBEED, INC
Warning

A161900-1G A161900-5G A161900-100G A161900-100MG A161900-10G A161900-250MG A161900-1G A161900-25G A161900-5G
103805-548EA 8 USD InStock
103805-548 103843-792 76783-888 76783-890 76783-892 76783-894 77319-806 77319-808 77319-810
2-Chloro-5-(trifluoromethyl)nicotinic acid 98%
2-Chloro-5-(trifluoromethyl)nicotinic acid
Precaución: for Research Use Only


Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 301.5 °C (760 mm Hg)
Melting Pt: 281 °C
Flash Pt: 136.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08741353
CAS Number: 505084-59-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty