Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluoro-3-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A206221-1G A206221-5G A206221-250MG A206221-1G A206221-10G
103817-694EA 13 USD InStock
103817-694 103848-364 76975-360 76975-362 77086-610 77671-824
2-Chloro-5-fluoro-3-methylpyridine 98%
2-Chloro-5-fluoro-3-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅ClFN
MW: 145.56
Boiling Pt: 177.9 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 61.409 °C
Storage Temperature: Ambient
MDL Number: MFCD04972725
CAS Number: 38186-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty