Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluoroanisole 97%

Supplier: AMBEED, INC
Warning

A225010-1G A225010-5G A225010-1G A225010-25G A225010-5G A225010-10G
103819-148EA 9 USD InStock
103819-148 103850-042 77088-710 77088-712 77088-714 77261-390
2-Chloro-5-fluoroanisole 97%
2-Chloro-5-fluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆ClFO
MW: 160.58 g/mol
Boiling Pt: 195
Storage Temperature: Ambient
MDL Number: MFCD00042573
CAS Number: 450-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty