Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-4-fluoro-2-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A183046-1G A183046-5G A183046-100G A183046-10G A183046-500G A183046-1G A183046-25G A183046-5G
103815-596EA 5 USD InStock
103815-596 103846-160 77287-326 77287-328 77287-330 77301-812 77301-814 77301-816
1-Chloro-4-fluoro-2-nitrobenzene 98%
1-Chloro-4-fluoro-2-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
MW: 175.55
Boiling Pt: 238.5
Melting Pt: 37…40 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00084853
CAS Number: 345-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty