Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-iodopyrazine 95%

Supplier: AMBEED, INC
Warning

A175503-1G A175503-100MG A175503-250MG A175503-1G
103807-048EA 41 USD InStock
103807-048 76784-422 76784-424 77082-698
2-Chloro-5-iodopyrazine 95%
2-Chloro-5-iodopyrazine
Precaución: for Research Use Only


Formula: C₄H₂ClIN₂
MW: 240.4300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09909701
CAS Number: 1057216-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty