Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxypyridine 98%

Supplier: AMBEED, INC
Warning

A137591-1G A137591-10G A137591-250MG A137591-25G A137591-1G A137591-5G
103802-476EA 106 USD InStock
103802-476 77149-912 77149-914 77298-912 77315-446 77315-448
2-Chloro-5-methoxypyridine 98%
2-Chloro-5-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₆ClNO
MW: 143.571 g/mol
Boiling Pt: 213.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06659513
CAS Number: 139585-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty