Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-phenylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A184055-1G A184055-5G A184055-100MG A184055-250MG
103815-706EA 54 USD InStock
103815-706 103846-266 77260-336 77260-338
2-Chloro-5-phenylpyrimidine 97%
2-Chloro-5-phenylpyrimidine
Precaución: for Research Use Only


Formula: C₁₀H₇ClN₂
MW: 190.629 g/mol
Boiling Pt: 369.9 °C (760 mm Hg)
Melting Pt: 131…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04035567
CAS Number: 22536-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty