Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine 95%

Supplier: AMBEED, INC
Warning

A632503-1G A632503-5G A632503-100MG A632503-250MG
103840-282EA 271 USD InStock
103840-282 76844-458 76844-460 76980-680 76980-682
1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine 95%
1-(6-Chloro-2-pyrazinyl)-4-methylpiperazine
Precaución: For research use only


Formula: C₉H₁₃ClN₄
MW: 212.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024895
CAS Number: 61655-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty