Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(trifluoromethyl)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A133112-1G A133112-10G A133112-1G A133112-25G A133112-5G
103801-828EA 11 USD InStock
103801-828 76811-332 77314-432 77314-434 77314-436
2-Chloro-6-(trifluoromethyl)benzaldehyde 97%
2-Chloro-6-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₄ClF₃O
Boiling Pt: 213.3 °C (760 mm Hg)
Flash Pt: 82.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06660231
CAS Number: 60611-22-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty