Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-iodo-3-pyridinol 97%

Supplier: AMBEED, INC
Warning

A175505-1G A175505-5G A175505-100MG A175505-25G A175505-5G A175505-10G A175505-250MG
103807-050EA 8 USD InStock
103807-050 103845-358 76998-344 77082-700 77082-702 77671-494 77881-340
2-Chloro-6-iodo-3-pyridinol 97%
2-Chloro-6-iodo-3-pyridinol
Precaución: for Research Use Only


Formula: C₅H₃ClINO
Boiling Pt: 394.5 °C (760 mm Hg)
Melting Pt: 141…145 °C
Flash Pt: 192.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10566651
CAS Number: 185220-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty