Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methoxyquinoline 98%

Supplier: AMBEED, INC
Warning

A208944-1G A208944-5G A208944-10G A208944-250MG A208944-1G A208944-5G
103817-902EA 42 USD InStock
103817-902 103848-590 77012-894 77012-896 77086-924 77086-926
2-Chloro-6-methoxyquinoline 98%
2-Chloro-6-methoxyquinoline
Precaución: for Research Use Only


Formula: C₁₀H₈ClNO
MW: 193.63
Melting Pt: 108…109 °C
Flash Pt: 141.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00799212
CAS Number: 13676-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty