Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-nitropyridine 95%

Supplier: AMBEED, INC
Warning

A140152-1G A140152-100MG A140152-10G A140152-250MG A140152-1G A140152-25G
103802-838EA 50 USD InStock
103802-838 77258-754 77258-756 77258-758 77315-950 77315-952
2-Chloro-6-nitropyridine 95%
2-Chloro-6-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₃ClN₂O₂
MW: 158.54
Melting Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114207
CAS Number: 94166-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty