Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorobenzoxazole 97%

Supplier: AMBEED, INC
Warning

A194342-5G A194342-25G A194342-10G A194342-1G A194342-5G
103847-252EA 6 USD InStock
103847-252 77085-170 77111-910 77111-912 77302-406
2-Chlorobenzoxazole 97%
2-Chlorobenzoxazole
Precaución: for Research Use Only


Formula: C₇H₄ClNO
Boiling Pt: 201.5 °C (760 mm Hg)
Melting Pt: 7 °C
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005766
CAS Number: 615-18-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty