Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroisonicotinonitrile 98%

Supplier: AMBEED, INC
Warning

A136532-100G A136532-10G A136532-500G A136532-25G A136532-5G
77149-872EA 44 USD InStock
77149-872 77149-874 77149-876 77315-218 77315-220
2-Chloroisonicotinonitrile 98%
2-Chloroisonicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₃ClN₂
Boiling Pt: 218.9 °C (760 mm Hg)
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00174318
CAS Number: 33252-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty