Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroquinolin-6-ol 97%

Supplier: AMBEED, INC
Warning

A173631-1G A173631-5G A173631-10G A173631-250MG A173631-1G A173631-25G A173631-5G
103806-852EA 13 USD InStock
103806-852 103845-152 76872-508 76872-510 77082-410 77082-412 77082-414
2-Chloroquinolin-6-ol 97%
2-Chloroquinolin-6-ol
Precaución: for Research Use Only


Formula: C₉H₆ClNO
Boiling Pt: 342.1±22.0 °C (760 mm Hg)
Flash Pt: 160.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12405093
CAS Number: 577967-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty