Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorothiazole-5-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A162388-1G A162388-5G A162388-10G A162388-250MG A162388-1G A162388-25G A162388-5G
103805-612EA 14 USD InStock
103805-612 103843-862 77259-666 77259-668 77319-902 77319-904 77319-906
2-Chlorothiazole-5-carbaldehyde 98%
2-Chlorothiazole-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₄H₂ClNOS
MW: 147.58 g/mol
Boiling Pt: 278.6 °C (760 mm Hg)
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD01568717
CAS Number: 95453-58-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty