Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutylacetic acid 97%

Supplier: AMBEED, INC
Warning

A371089-1G A371089-5G A371089-10G A371089-250MG A371089-1G A371089-25G A371089-5G
103826-474EA 23 USD InStock
103826-474 103837-494 76694-334 76694-336 76840-742 77306-824 77306-826
Cyclobutylacetic acid 97%
Cyclobutylacetic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
Boiling Pt: 210.7 °C (760 mm Hg)
Flash Pt: 103.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411067
CAS Number: 6540-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty