Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ɑ-Cyclohexyl-DL-mandelic acid 95%

Supplier: AMBEED, INC
Warning

A101998-5G A101998-100G A101998-10G A101998-25G A101998-5G
103821-942EA 9 USD InStock
103821-942 77188-578 77188-580 77188-582 77282-888
ɑ-Cyclohexyl-DL-mandelic acid 95%
alpha-Cyclohexyl-DL-mandelic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₈O₃
MW: 234.30 g/mol
Boiling Pt: 408.6 °C (760 mm Hg)
Melting Pt: 163 °C
Flash Pt: 215.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00194543
CAS Number: 4335-77-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty