Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclohexylpropanoic acid 98%

Supplier: AMBEED, INC
Warning

A151505-1G A151505-5G A151505-10G A151505-1G A151505-25G A151505-5G
103804-262EA 28 USD InStock
103804-262 103842-414 77010-822 77317-980 77317-982 77317-984
2-Cyclohexylpropanoic acid 98%
2-Cyclohexylpropanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00268172
CAS Number: 6051-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty