Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Danger

A274547-1G A274547-1G A274547-25G A274547-5G A274547-10G
103813-200EA 6 USD InStock
103813-200 103853-128 77092-950 77092-952 77092-954 77276-844
Cyclopropylboronic acid pinacol ester 97%
Cyclopropylboronic acid pinacol ester


Formula: C₉H₁₇BO₂
Boiling Pt: 146 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty