Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclopropylaniline hydrochloride 95%

Supplier: AMBEED, INC
Warning

A271903-1G A271903-5G A271903-100MG A271903-5G
103813-074EA 103 USD InStock
103813-074 103852-986 77014-434 77092-778
2-Cyclopropylaniline hydrochloride 95%
2-Cyclopropylaniline hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD04109471
CAS Number: 1056454-83-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty