Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-4,6-dihydroxypyrimidine 95%

Supplier: AMBEED, INC
Warning

A107138-1G A107138-5G A107138-100MG A107138-10G A107138-1G A107138-5G
103797-132EA 22 USD InStock
103797-132 103822-892 77283-542 77283-544 77283-546 77309-758
2-Ethyl-4,6-dihydroxypyrimidine 95%
2-Ethyl-4,6-dihydroxypyrimidine
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09907955
CAS Number: 3709-98-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty