Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-9,10-anthraquinone 98%

Supplier: AMBEED, INC
Warning

A122554-1000G A122554-100G A122554-25G A122554-500G A122554-1000G
103808-252EA 81 USD InStock
103808-252 77026-522 77026-524 77026-526 77297-872
2-Ethyl-9,10-anthraquinone 98%
2-Ethyl-9,10-anthraquinone
Precaución: for Research Use Only


Formula: C₁₆H₁₂O₂
Boiling Pt: 180…190 °C (760 mm Hg)
Melting Pt: 108…111 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00001237
CAS Number: 84-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty