Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethynylpyrazine 97%

Supplier: AMBEED, INC
Warning

A242073-1G A242073-5G A242073-100MG A242073-250MG
103811-452EA 115 USD InStock
103811-452 103851-262 77153-104 77153-106
2-Ethynylpyrazine 97%
2-Ethynylpyrazine
Precaución: for Research Use Only


Formula: C₆H₄N₂
Boiling Pt: 180.3±20.0 °C (760 mm Hg)
Flash Pt: 73.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189804
CAS Number: 153800-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty