Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Fluoro-4'-methoxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A121223-100G A121223-25G A121223-10G
77271-506EA 86 USD InStock
77271-506 77271-508 77747-880
2'-Fluoro-4'-methoxyacetophenone 98%
2'-Fluoro-4'-methoxyacetophenone
Precaución: for Research Use Only


Formula: C₉H₉FO₂
MW: 168.17
Boiling Pt: 243
Melting Pt: 52…54 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042290
CAS Number: 74457-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty