Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-p-toluonitrile 98%

Supplier: AMBEED, INC
Danger

A154715-1G A154715-5G A154715-10G A154715-1G A154715-25G A154715-5G
103804-672EA 9 USD InStock
103804-672 103842-864 76768-286 77300-072 77300-074 77318-604
2-Fluoro-p-toluonitrile 98%
2-Fluoro-p-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆FN
MW: 135.14
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD03094311
CAS Number: 85070-67-3
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty