Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(trifluoromethoxy)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A372483-1G A372483-25G A372483-5G A372483-10G A372483-250MG
103826-504EA 21 USD InStock
103826-504 103837-522 76840-778 76840-780 76840-782 77264-114 77264-116
2-Fluoro-5-(trifluoromethoxy)benzoic acid 97%
2-Fluoro-5-(trifluoromethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Boiling Pt: 251.9 °C (760 mm Hg)
Flash Pt: 106.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04115887
CAS Number: 886497-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty