Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-nitrobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A232987-5G A232987-25G A232987-100G A232987-10G A232987-5G A232987-500G
103850-622EA 6 USD InStock
103850-622 77089-566 77275-778 77275-780 77303-950 77748-399
2-Fluoro-5-nitrobenzoic acid 98%
2-Fluoro-5-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 142…144 °C
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00134238
CAS Number: 7304-32-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty