Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Danger

A841124-5G A841124-100G A841124-500G
103841-206EA 12 USD InStock
103841-206 76845-446 77293-966 77293-968
2-Fluoro-6-(trifluoromethyl)pyridine 98%
2-Fluoro-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₃F₄N
Boiling Pt: 241.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03092901
CAS Number: 94239-04-0
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty