Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A600677-25G A600677-100G A600677-10G A600677-500G
103840-114EA 5 USD InStock
103840-114 76844-274 76844-276 77104-022 77104-024 77104-026
2-Fluorobenzyl alcohol 97%
2-Fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇FO
MW: 126.13
Boiling Pt: 200
Melting Pt: 91…94 °C (760 mm Hg)
Density: 1.17
Storage Temperature: Ambient
MDL Number: MFCD00004601
CAS Number: 446-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty