Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorocyclopropanecarboxylic acid 97%

Supplier: AMBEED, INC
Danger

A129518-1G A129518-100MG A129518-250MG A129518-1G
103801-294EA 41 USD InStock
103801-294 76971-998 76972-000 77313-676
2-Fluorocyclopropanecarboxylic acid 97%
2-Fluorocyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₅FO₂
MW: 104.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21604147
CAS Number: 156816-78-3
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty