Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Acetonedicarboxylic acid anhydride 97%

Supplier: AMBEED, INC
Warning

A139439-1G A139439-100MG A139439-250MG A139439-50MG
103802-744EA 509 USD InStock
103802-744 77190-332 77190-334 77190-336
1,3-Acetonedicarboxylic acid anhydride 97%
1,3-Acetonedicarboxylic acid anhydride
Precaución: for Research Use Only


Formula: C₅H₄O₄
Boiling Pt: 354.9 °C (760 mm Hg)
Melting Pt: 138 °C
Flash Pt: 165.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783094
CAS Number: 10521-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty